Desiccators
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
65231 65238 65243
BRND65231EA 242 GBP
BRND65231 BRND65238 BRND65243
Desiccators
Desiccators Desiccators, Glass
Borosilicate glass 3.3 with precisely ground flat flanges.
  • Base ring surface is studded to improve stability and help prevent cracks
  • Interchangeable lid
Desiccators with sockets are supplied with 24/29 socket and interchangeable stopcock.
Certifications: DIN 12491