Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2.36853 2.36845 2.36851 2.36843 2.36838 2.36848 2.36850 2.36854 2.36842 2.36852 2.36849 2.36847 2.36846
BRND2.36853EA 59.7 GBP
BRND2.36853 BRND2.36845 BRND2.36851 BRND2.36843 BRND2.36838 BRND2.36848 BRND2.36850 BRND2.36854 BRND2.36842 BRND2.36852 BRND2.36849 BRND2.36847 BRND2.36846
Volumetric flasks, class A, amber markings, BLAUBRAND® ETERNA
Flasks Volumetric Flasks
Borosilicate glass 3.3, with PP stopper.
  • Calibrated to contain (TC, In)
  • Mark and inscriptions in ETERNA amber stain
  • DE-M marking
  • Total order from one batch, supplied with batch certificate
*Wide neck
Certifications: DIN EN ISO 1042
Ordering information: Delivered with batch number and enclosed one batch certificate per packing unit of the manufacturer. On demand also available with individual certificate or DAkkS calibration certificate.