Watch glasses
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
150010 150015 150020 150030 150040 150045 150050 150060 150065 150070 150080
BRND150010EA 28.3 GBP
BRND150010 BRND150015 BRND150020 BRND150030 BRND150040 BRND150045 BRND150050 BRND150060 BRND150065 BRND150070 BRND150080
Watch glasses
Watch Glasses
Soda-lime glass.
  • Good chemical and physical properties
  • Use in applications that do not involve high thermal stresses
  • Quick and easy to clean
With fire-polished edges. Stress-reduced to minimise risk of shattering.
Certifications: DIN 12341.