Stoppers for conical ground joints
Supplier: Brand
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
144515 144520 144525 144530 144535 144540 144545
BRND144515EA 9.9 GBP
BRND144515 BRND144520 BRND144525 BRND144530 BRND144535 BRND144540 BRND144545
Stoppers for conical ground joints
Stoppers Glass Stoppers
Boro 3.3.
  • Solid or semi-hollow design, with octagonal handle
Certifications: DIN 12252