Lockout Visit Bags
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
830931 830933 830934
BRDY830931EA 199.5 GBP
BRDY830931 BRDY830933 BRDY830934
Lockout Visit Bags
Bags Storage Bags
These Lockout visit bag has several pockets to store and smoothly retrieve any lockout/tagout product.
A small, complimentary bag to store accessories and small devices accompanies the Lockout/Tagout Visit Bag. All padlocks and keys, hasps or circuit breakers go into the small bag.The small bag fits perfectly in one of the big bag’s pockets and can be used as a standalone or in combination with the Lockout/Tagout Visit Bag.
Some of the bigger pockets are excellent storage compartments for the larger gate valve lockout device, the universal lockout device and similar size products.