Padlocks, brass, with steel shackle
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
805831 805832 805834 805827 805828 805829 805830
BRDY805831EA 45.08 GBP
BRDY805831 BRDY805832 BRDY805834 BRDY805827 BRDY805828 BRDY805829 BRDY805830
Padlocks, brass, with steel shackle
Security Locks
These durable padlocks feature solid brass bodies and stainless steel or hardened steel shackles that resist corrosion and sparks.
  • Percision pin tumbler cylinder
  • Two keys supplied with each lock