Label printer, BMP™41
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
133254 711213
BRDY133254EA 545 GBP
BRDY133254 BRDY711213
Label printer, BMP™41
Printers Label Printers
The BMP™41 portable label printer creates die-cut or continuous labels anywhere and at anytime. It can produce up to 25 mm wide labels which can be used as multi area labels mostly on labelling datacom, wires, cables and audio/visual equipments.
Delivery information: Supplied with nickel battery, MC1-1000-595-WT-BK cartridge, USB cable, AC adapter, power cord, product CD, brady printer drivers CD and quick start guide.