ISO safety signs - Wear laboratory coat, self adhesive
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
311364 311365 311366 311367 311368 311373 311374 311375 311376 311377 311378 311379
BRDY311364EA 5.14 GBP
BRDY311364 BRDY311365 BRDY311366 BRDY311367 BRDY311368 BRDY311373 BRDY311374 BRDY311375 BRDY311376 BRDY311377 BRDY311378 BRDY311379
ISO safety signs - Wear laboratory coat, self adhesive
Signs
Safety communication is vital in workplace accident prevention. Instructing and reminding employees and visitors that a laboratory coat must be worn is of crucial importance.
  • Increase safety using internationally recognised signs
  • Compliant with ISO 7010/ISO 3864
  • Designs are available in multiple top selling materials
  • Polyester over-laminated with a clear polyester
  • Permanent acrylic adhesive
Laboratory coats are a critical component of worker protection in both laboratory and clinical/patient care areas. In order to protect against accidental spill orMore Product Information