SPC® Absorbents SitePoint Mobile Unit, Empty
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
309075
BRDY309075EA 374.63 GBP
BRDY309075
SPC® Absorbents SitePoint Mobile Unit, Empty
Spill Control Materials
Spill kit.
  • Features two, 200 mm wheels for mobility, and anchor points for fixed installation
  • Highly customisable
  • Fix items to backboard using self-tapping screws or bolts
  • Suitable for inside or outdoor use
  • Fully recyclable and chemical resistant
Ideal to move spill control equipment near specific risks. Features two rubber wheels for easy trolley movement to wherever it is required. Perfect during maintenance activities, at construction sites and other temporary locations. Includes three anchor points on the base, allowing the unit to be bolted toMore Product Information