Label printer BBP™12
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
195566 195812 195567 195966 198597 198600 195967
116-0605EA 940 GBP
116-0605 116-0609 116-0606 BRDY195966 BRDY198597 BRDY198600 BRDY195967
Label printer BBP™12
Printers Label Printers
The BBP™12 bench top label printer is a compact, reliable and affordable entry level printer to identify cables, components, products and laboratory samples for customers who require low volumes of various, quality identification labels. The printer can print on a multitude of materials in various sizes including nylon cloth, paper, polyester, polyimides, polyolefin, polypropylene, tamper evident materials, vinyl (also self-laminating), vinyl cloth, to produce labels, tags, cable markers, sleeves (both ladder and continuous).
  • Accurate image printing, 300 dpi gives good print resolution
More Product Information