Code Corporation Barcode Reader Accessories
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
176703 176701 176702 176700
BRDY176703EA 28.56 GBP
BRDY176703 BRDY176701 BRDY176702 BRDY176700
Code Corporation Barcode Reader Accessories
Barcode and RFID Systems
Accessories available for barcode readers.
Use the 1,8 m (BRDY176701) or 2,4 m (BRDY176700) coiled RS232 Affinity® cable CRA-C511 with code's cabled barcode readers.
Model BRDY176702 code power supply is an international power supply with adapter clips for the US, EU, UK and AU (barrel plug, 12 V/3,3 A).
Model BRDY176703 code power supply is an international power supply with USB connection.
Order Now
SPECIFICATIONS
Environmentally Preferable