Self-Laminating Vinyl Wrap-Around Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
170740 170743 170862 170750 170745 170861 170863 170741 170742 170744 170746 170747 170748 170749 170751 170752 170753 170754 170755 170756 170757 170758 170759 170760 170761 170864 170865 170866 170867 170868 170869 170870
BRDY170740EA 77.59 GBP
BRDY170740 BRDY170743 BRDY170862 BRDY170750 BRDY170745 BRDY170861 BRDY170863 BRDY170741 BRDY170742 BRDY170744 BRDY170746 BRDY170747 BRDY170748 BRDY170749 BRDY170751 BRDY170752 BRDY170753 BRDY170754 BRDY170755 BRDY170756 BRDY170757 BRDY170758 BRDY170759 BRDY170760 BRDY170761 BRDY170864 BRDY170865 BRDY170866 BRDY170867 BRDY170868 BRDY170869 BRDY170870
Self-Laminating Vinyl Wrap-Around Labels for M410, BMP41, M510, BMP51, BMP53 and M511 Printers
Labels
Mark cables and wires with the B-427 self-laminating vinyl label material, designed with a clear self-laminating protective layer. Print almost anywhere with a durable Brady portable label printer.
  • For use on both smooth and textured/rough surfaces
  • Self-extinguishing and resistant to abrasion, solvents, oil, grease, weather and extreme temperatures
  • Permanent acrylic adhesive works great on smooth, rough, powder-coated, and irregular surfaces
  • Made with a transparent, non-printable overlaminate for increased protection
Flexible labels are designed forMore Product Information