Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
170859
BRDY170859EA 63.14 GBP
BRDY170859
Aggressive Adhesive Multi-Purpose Polyester Labels with Ribbon for M510, BMP51, BMP53 and M511 Printers
Labels
Easily label components with the thermal transfer, glossy permanent polyester label material (B-422). Print almost anywhere with a durable Brady handheld label printer.
  • Versatile - Designed for a wide range of applications such as labeling equipment, circuit boards, panel components and barcodes
  • Tough - Constructed to withstand abrasions, extreme temperatures and solvents such as alcohol, oil and water
  • Adhesive - Made with a permanent acrylic adhesive that sticks to rough, smooth and powder-coated flat surfaces
Ideal for asset tracking, barcodeMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable