Labelling tapes, dissolvable paper tape for thermal transfer printers
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
150603 150604 150605 150606 150607
BRDY150603EA 35.81 GBP
BRDY150603 BRDY150604 BRDY150605 BRDY150606 BRDY150607
Labelling tapes, dissolvable paper tape for thermal transfer printers
Labels
Continuous tapes for printers.
  • Removable acrylic adhesive
  • Matte finish
  • Autoclavable
  • Dissolves completely in water leaving no adhesive residue
  • Continuous tape is not cut to length, it gets cut by the printer
  • This node contains only supplies sold with the ribbon seperate, it is not in the same enclosure as the tape/sleeves
Water dissolvable paper labels are self-adhesive and extremely easy to apply. Exposing the label to warm water will completely dissolve it within 30 seconds, without damage to the surface it was applied to, and without leaving anyMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable