Mop systems, HandySorb
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
150597 150598
BRDY150597EA 297.59 GBP
BRDY150597 BRDY150598
Mop systems, HandySorb
Mops
HandySorb is a unique and practical tool to safely remove spills in a highly efficient and easy way.
Safe spill clean-up: works with inert pads or pillows that can be replaced without ever touching the absorbed fluids
Easy to use: offers 1,83 m reach when fully extended and features a no touch snap-on system for pads
Highly efficient: each pad absorbs in seconds up to 25 times its weight