Labels for BBP33 printers, type B-422
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
133815 133822 133826 133886 133957 133973 134003 134004 133768 133797 133800
BRDY133815EA 100.04 GBP
BRDY133815 BRDY133822 BRDY133826 BRDY133886 BRDY133957 BRDY133973 BRDY134003 BRDY134004 BRDY133768 BRDY133797 BRDY133800
Labels for BBP33 printers, type B-422
Labels
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
  • Designed to withstand exposure to numerous solvents
  • Service temperature of –40 to +100 °C
Aggressive adhesive good for powder-coated and textured flat surfaces including low surface energy plastics.
Certifications: UL approved; CSA accepted.