Ribbons for PR200 plus PR400 plus and PR600 plus printers, R-4300 series
Supplier: Brady
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
011510 800732 800731 035243 035244 011511 035268 035246 035269
BRDY011510EA 72.55 GBP
BRDY011510 BRDY800732 BRDY800731 BRDY035243 BRDY035244 BRDY011511 BRDY035268 BRDY035246 BRDY035269
Ribbons for PR200 plus PR400 plus and PR600 plus printers, R-4300 series
Printer Ribbons
Ribbon, with strong strong environmental resistance against smearing and chemicals, and with reliable, sharp-edge print quality to maximise bar code readability.