Insulin syringes, Omnifix® 100 Solo and Duo
Supplier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
9161376V 9161708V
720-2558EA 66.7 GBP
720-2558 BRAU9161708V
Insulin syringes, Omnifix® 100 Solo and Duo
Syringes Syringes, Medical
PP, three-piece syringe for injection of U-100 insulin (50 or 100 IU/ml) for veterinary use. Smooth action, natural latex-free plunger stoppers with double sealing ring.
  • Permanent, easy-to-read graduations
  • Safe plunger backstop
  • Centric catheter filling
Packaging: 100 units per box.