Tubings, Original Perfusor®
Supplier: B. Braun
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8722935 8255172 8255059 8255253 8722919 8723060 8255067 8272565
720-2584EA 238 GBP
720-2584 BRAU8255172 BRAU8255059 BRAU8255253 BRAU8722919 BRAU8723060 BRAU8255067 BRAU8272565
Tubings, Original Perfusor®
Infusions
Original-Perfusor tubings for use with all syringe pumps inferior to four-bar pressure.
  • Minimized priming volumes
  • Luer-Lock fitting
  • Injection filter 0,22 μm and back-check valve for parallel infusions