Anti-Digoxin Mouse Monoclonal Antibody [clone: 2E2] (ALEXA FLUOR® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BSM-0356M-A750
BOSSBSM-0356M-A750EA 1030 GBP
BOSSBSM-0356M-A750
Anti-Digoxin Mouse Monoclonal Antibody [clone: 2E2] (ALEXA FLUOR® 750)
Antibodies
Digoxin is a beta blocker drug originally derived from the foxglove plant, Digitalis lanata. Digoxin is used primarily to improve the pumping ability of the heart in congestive heart failure (CHF), and treat problems such as high blood pressure. It is also used to help normalise some dysrhythmias (abnormal types of heartbeat).

Type: Primary
Antigen: Digoxin
Clonality: Monoclonal
Clone: 2E2
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range:
Host: Mouse
Isotype: IgG
Reactivity: Others
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR