Anti-Ube2N Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-8351R-A680
BOSSBS-8351R-A680EA 838 GBP
BOSSBS-8351R-A680
Anti-Ube2N Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Ubiquitination is an important mechanism through which three classes of enzymes act in concert to target short-lived or abnormal proteins for destruction. The three classes of enzymes involved in ubiquitination are the ubiquitin-activating enzymes (E1s), the ubiquitin-conjugating enzymes (E2s) and the ubiquitin-protein ligases (E3s). UBC13, also known as UBE2N or BLU, is a 152 amino acid member of the E2 ubiquitin-conjugating enzyme family. Existing as a heterodimer with Mms2 (also known as UBE2V2), UBC13 catalyzes the ATP-dependent synthesis of non-canonical polyubiquitin chains, a processMore Product Information