Anti-CA11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-8244R-A555
BOSSBS-8244R-A555EA 838 GBP
BOSSBS-8244R-A555
Anti-CA11 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
Antibodies
Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes responsible for catalyzing the reversible hydration of carbon dioxide. CAs show extensive diversity in their distribution and subcellular localization. They are involved in a variety of biological processes, including calcification, bone resorption, respiration, acid-base balance and the formation of aqueous humor, saliva, gastric juice and cerebrospinal fluid. CA XI, also referred to as carbonic anhydrase-related protein 11 precursor (CA-RP XI) or carbonic anhydrase-related protein 2 (CA-RP II), is a member ofMore Product Information