Anti-GNL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-8040R-A680
BOSSBS-8040R-A680EA 838 GBP
BOSSBS-8040R-A680
Anti-GNL1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
The GNL1 gene, identified in the human major histocompatibility complex class I region, shows a high degree of similarity with its mouse counterpart. The GNL1 gene is located less than 2 kb centromeric to HLA-E, in the same transcriptional orientation. GNL1 is telomeric to HLA-B and HLA-C.

Type: Primary
Antigen: GNL1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 301-400/607
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR