Anti-ABHD14B Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7945R-CY5.5
BOSSBS-7945R-CY5.5EA 844 GBP
BOSSBS-7945R-CY5.5
Anti-ABHD14B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
ABHD14B has hydrolase activity towards p-nitrophenyl butyrate (in vitro). It may activate transcription. There are 2 isoforms produced by alternative splicing.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: ABHD14B
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR