Anti-CLK3 Rabbit Polyclonal Antibody (Cy5.5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7908R-CY5.5
BOSSBS-7908R-CY5.5EA 844 GBP
BOSSBS-7908R-CY5.5
Anti-CLK3 Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Phosphorylates serine- and arginine-rich (SR) proteins of the spliceosomal complex. May be a constituent of a network of regulatory mechanisms that enable SR proteins to control RNA splicing. Phosphorylates serines, threonines and tyrosines.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: CLK3
Clonality: Polyclonal
Clone:
Conjugation: Cy5.5®
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR