Anti-RAB35 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7765R-A350
BOSSBS-7765R-A350EA 838 GBP
BOSSBS-7765R-A350
Anti-RAB35 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
In the process of endocytosis, essential rate-limiting regulator of a fast recycling pathway back to the plasma membrane. During cytokinesis, required for the postfurrowing terminal steps, namely for intercellular bridge stability and abscission, possibly by controlling phosphatidylinositol 4,5-bis phosphate (PIP2) and SEPT2 localization at the intercellular bridge.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: RAB35
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR