Anti-ASB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-7736R-A350
BOSSBS-7736R-A350EA 838 GBP
BOSSBS-7736R-A350
Anti-ASB3 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Members of the suppressor of cytokine signaling (SOCS) family of proteins contain C-terminal regions of homology called the SOCS box, which serves to couple SOCS proteins and their binding partners with the elongin B and C complex. Serveral other families of proteins also contain SOCS boxes but differ from the SOCS proteins in the type of domain they contained upstream of the SOCS box. Four members of the ankyrin repeat and SOCS box-containing (ASB) protein family are identified and termed as ASB-1, ASB-2, ASB-3 and ASB-4. ASB-1 is expressed in multiple organs, including the hematopoieticMore Product Information
Order Now

SPECIFICATIONS

Antigen symbol ASB3
Antigen name Ankyrin Repeat And SOCS Box Containing 3
Antigen synonyms ASB3_HUMAN|Weakly similar to ankyrin brain variant 2.|ASB-3|Ankyrin repeat and SOCS box containing 3|ASB3
Antibody type Primary
Clonality Polyclonal
Conjugation Alexa Fluor® 350
Reactivity Human,Rat,Mouse
Host Rabbit
Gene ID 51130
Isotype IgG
ImmunoChemistry Yes
Form liquid
Purification Purified by Protein A
Concentration 1 μg/μl
Cross adsorption No
Storage buffer Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
Storage temperature Store at –20 °C for 12 months
Shipping temperature 4°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR