Anti-CCAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-7064R-A680
BOSSBS-7064R-A680EA 838 GBP
BOSSBS-7064R-A680
Anti-CCAR1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
Associates with components of the Mediator and p160 coactivator complexes that play a role as intermediaries transducing regulatory signals from upstream transcriptional activator proteins to basal transcription machinery at the core promoter. Recruited to endogenous nuclear receptor target genes in response to the appropriate hormone. Also functions as a p53 coactivator. May thus play an important role in transcriptional regulation (By similarity). May be involved in apoptosis signaling in the presence of the reinoid CD437. Apoptosis induction involves sequestration of 14-3-3 protein(s) andMore Product Information