Anti-Calpastatin Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6701R-A750
BOSSBS-6701R-A750EA 858 GBP
BOSSBS-6701R-A750
Anti-Calpastatin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Specific inhibition of calpain (calcium-dependent cysteine protease). Plays a key role in postmortem tenderisation of meat and have been proposed to be involved in muscle protein degradation in living tissue.

Type: Primary
Antigen: Calpastatin
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 431-530/708
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR