Anti-GIRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6672R-A750
BOSSBS-6672R-A750EA 858 GBP
BOSSBS-6672R-A750
Anti-GIRK1 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
This potassium channel is controlled by G proteins. Inward rectifier potassium channels are characterised by a greater tendency to allow potassium to flow into the cell rather than out of it. Their voltage dependence is regulated by the concentration of extracellular potassium; as external potassium is raised, the voltage range of the channel opening shifts to more positive voltages. The inward rectification is mainly due to the blockage of outward current by internal magnesium. This receptor plays a crucial role in regulating the heartbeat.

Type: Primary
Antigen: GIRK1
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR