Anti-ALOX15B Rabbit Polyclonal Antibody (Cy5.5®)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-6335R-CY5.5
BOSSBS-6335R-CY5.5EA 844 GBP
BOSSBS-6335R-CY5.5
Anti-ALOX15B Rabbit Polyclonal Antibody (Cy5.5®)
Antibodies
Non-heme iron-containing dioxygenase that catalyzes the stereo-specific peroxidation of free and esterified polyunsaturated fatty acids generating a spectrum of bioactive lipid mediators. Converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid/(15S)-HPETE. Also acts on linoleic acid to produce 13-hydroxyoctadecadienoic acid/13-HPODE. Has no detectable 8S-lipoxygenase activity but reacts with (8S)-HPETE to produce (8S,15S)-diHPETE. May regulate progression through the cell cycle and cell proliferation. May also regulate cytokine secretion by macrophages and therefore play a role inMore Product Information