Anti-LCMT1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-6249R-CY5
BOSSBS-6249R-CY5EA 844 GBP
BOSSBS-6249R-CY5
Anti-LCMT1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
LCMT1 catalyzes the methylation of the carboxyl group of the C-terminal leucine residue (leu309) of the catalytic subunit of protein phosphatase-2A (PPP2CA; MIM 176915)

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: LCMT1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR