Anti-MAD2L1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5731R-A647
BOSSBS-5731R-A647EA 838 GBP
BOSSBS-5731R-A647
Anti-MAD2L1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Mad2L1 is required for the execution of the mitotic checkpoint which monitors the process of kinetochore spindle attachment and delays the onset of anaphase when this process is not complete. It inhibits the activity of the anaphase promoting complex by sequestering CDC20 until all chromosomes are aligned at the metaphase plate.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MAD2L1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR