Anti-ILK-1 Thr173 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5443R-A750
BOSSBS-5443R-A750EA 864 GBP
BOSSBS-5443R-A750
Anti-ILK-1 Thr173 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Receptor-proximal protein kinase regulating integrin-mediated signal transduction. May act as a mediator of inside-out integrin signaling. Focal adhesion protein part of the complex ILK-PINCH. This complex is considered to be one of the convergence points of integrin- and growth factor-signaling pathway. Could be implicated in mediating cell architecture, adhesion to integrin substrates and anchorage-dependent growth in epithelial cells. Phosphorylates beta-1 and beta-3 integrin subunit on serine and threonine residues, but also AKT1 and GSK3B.

Type: Primary
Antigen: ILK-1 Thr173
Clonality:More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR