Anti-PON1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-5107R-A647
BOSSBS-5107R-A647EA 838 GBP
BOSSBS-5107R-A647
Anti-PON1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Antibodies
Hydrolyzes the toxic metabolites of a variety of organophosphorus insecticides. Capable of hydrolyzing a broad spectrum of organophosphate substrates and lactones, and a number of aromatic carboxylic acid esters. Mediates an enzymatic protection of low density lipoproteins against oxidative modification and the consequent series of events leading to atheroma formation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PON1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR