Anti-PCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4972R-A488
BOSSBS-4972R-A488EA 838 GBP
BOSSBS-4972R-A488
Anti-PCK1 Rabbit Polyclonal Antibody (Alexa Fluor® 488)
Antibodies
PCK1 is a main control point for the regulation of gluconeogenesis. This cytosolic enzyme, along with GTP, catalyzes the formation of phosphoenolpyruvate from oxaloacetate, with the release of carbon dioxide and GDP. The expression of the corresponding gene can be regulated by insulin, glucocorticoids, glucagon, cAMP, and diet. A mitochondrial isozyme has also been characterized.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: PCK1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 488
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR