Anti-VWF Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4754R-CY7
BOSSBS-4754R-CY7EA 844 GBP
BOSSBS-4754R-CY7
Anti-VWF Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Important in the maintenance of hemostasis, it promotes adhesion of platelets to the sites of vascular injury by forming a molecular bridge between sub-endothelial collagen matrix and platelet-surface receptor complex GPIb-IX-V. Also acts as a chaperone for coagulation factor VIII, delivering it to the site of injury, stabilizing its heterodimeric structure and protecting it from premature clearance from plasma.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VWF
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR