Anti-ATP receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-4065R-A750
BOSSBS-4065R-A750EA 835 GBP
BOSSBS-4065R-A750
Anti-ATP receptor Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Ion channel gated by extracellular ATP involved in a variety of cellular responses, such as excitatory postsynaptic responses in sensory neurons, neuromuscular junctions (NMJ) formation, hearing, perception of taste and peristalsis. In the inner ear, regulates sound transduction and auditory neurotransmission, outer hair cell electromotility, inner ear gap junctions, and K(+) recycling. Mediates synaptic transmission between neurons and from neurons to smooth muscle.

Type: Primary
Antigen: ATP receptor
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 251-350/4More Product Information
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR