Anti-IDH3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3946R-HRP
BOSSBS-3946R-HRPEA 853 GBP
BOSSBS-3946R-HRP
Anti-IDH3A Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antibodies
Isocitrate dehydrogenases catalyze the oxidative decarboxylation of isocitrate to 2-oxoglutarate. These enzymes belong to two distinct subclasses, one of which utilizes NAD(+) as the electron acceptor and the other NADP(+). Five isocitrate dehydrogenases have been reported: three NAD(+)-dependent isocitrate dehydrogenases, which localize to the mitochondrial matrix, and two NADP(+)-dependent isocitrate dehydrogenases, one of which is mitochondrial and the other predominantly cytosolic. NAD(+)-dependent isocitrate dehydrogenases catalyze the allosterically regulated rate-limiting step of theMore Product Information