Anti-Cubilin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3901R-A750
BOSSBS-3901R-A750EA 835 GBP
BOSSBS-3901R-A750
Anti-Cubilin Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Cotransporter which plays a role in lipoprotein, vitamin and iron metabolism, by facilitating their uptake. Binds to ALB, MB, Kappa and lambda-light chains, TF, hemoglobin, GC, SCGB1A1, APOA1, high density lipoprotein, and the GIF-cobalamin complex. The binding of all ligands requires calcium. Serves as important transporter in several absorptive epithelia, including intestine, renal proximal tubules and embryonic yolk sac. Interaction with LRP2 mediates its trafficking throughout vesicles and facilitates the uptake of specific ligands like GC, hemoglobin, ALB, TF and SCGB1A1. InteractionMore Product Information