Anti-HSD17B2 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3856R-CY7
BOSSBS-3856R-CY7EA 844 GBP
BOSSBS-3856R-CY7
Anti-HSD17B2 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
Capable of catalyzing the interconversion of testosterone and androstenedione, as well as estradiol and estrone. Also has 20-alpha-HSD activity. Uses NADH while EDH17B3 uses NADPH.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: HSD17B2
Clonality: Polyclonal
Clone:
Conjugation: Cy7®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR