Anti-ST6GAL1 Rabbit Polyclonal Antibody (Cy7®)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3793R-CY7
BOSSBS-3793R-CY7EA 844 GBP
BOSSBS-3793R-CY7
Anti-ST6GAL1 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene is a type II membrane protein that catalyzes the transfer of sialic acid from CMP sialic acid to galactose-containing substrates. The encoded protein, which is normally found in the Golgi apparatus but which can be proteolytically processed to a soluble form, is involved in the generation of the cell surface carbohydrate determinants and differentiation antigens HB6, CDw75, and CD76. This protein is a member of glycosyltransferase family 29. Three transcript variants encoding two different isoforms have been found for this gene.
Recommended Dilutions: IF(More Product Information