Anti-CRTC1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-3453R-A350
BOSSBS-3453R-A350EA 864 GBP
BOSSBS-3453R-A350
Anti-CRTC1 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
Antibodies
Transcriptional coactivator for CREB1 which activates transcription through both consensus and variant cAMP response element (CRE) sites. Acts as a coactivator, in the SIK/TORC signaling pathway, being active when dephosphorylated and acts independently of CREB1 'Ser-133' phosphorylation. Enhances the interaction of CREB1 with TAF4. Regulates the expression of specific CREB-activated genes such as the steroidogenic gene, StAR. Potent coactivator of PGC1alpha and inducer of mitochondrial biogenesis in muscle cells. Also coactivator for TAX activation of the human T-cell leukemia virus type 1 (More Product Information