Anti-MST1 Rabbit Polyclonal Antibody (Cy5®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-3294R-CY5
BOSSBS-3294R-CY5EA 864 GBP
BOSSBS-3294R-CY5
Anti-MST1 Rabbit Polyclonal Antibody (Cy5®)
Antibodies
Protein which catalyzes the phosphorylation of serine or threonine residues on target proteins by using ATP as phosphate donor. Such phosphorylation may cause changes in the function of the target protein. Protein kinases share a conserved catalytic core common to both serine/ threonine and tyrosine protein kinases.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: MST1
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR