Anti-LTK Rabbit Polyclonal Antibody (HRP)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-15500R-HRP
BOSSBS-15500R-HRPEA 844 GBP
BOSSBS-15500R-HRP
Anti-LTK Rabbit Polyclonal Antibody (HRP)
Antibodies
LTK is a member of the ros/insulin receptor family of tyrosine kinases. Tyrosine-specific phosphorylation of proteins is a key to the control of diverse pathways leading to cell growth and differentiation. Multiple transcript variants encoding different isoforms have been found for this gene.

Type: Primary
Antigen: LTK
Clonality: Polyclonal
Clone:
Conjugation: HRP
Public Immunogen Range: 41-140/864
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR