Anti-C20orf14 Rabbit Polyclonal Antibody (Cy7®)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-15094R-CY7
BOSSBS-15094R-CY7EA 844 GBP
BOSSBS-15094R-CY7
Anti-C20orf14 Rabbit Polyclonal Antibody (Cy7®)
Antibodies
The protein encoded by this gene appears to be involved in pre-mRNA splicing, possibly acting as a bridging factor between U5 and U4/U6 snRNPs in formation of the spliceosome. The encoded protein also can bind androgen receptor, providing a link between transcriptional activation and splicing.

Type: Primary
Antigen: C20orf14
Clonality: Polyclonal
Clone:
Conjugation: Cy7
Public Immunogen Range: 331-430/941
Host: Rabbit
Isotype: IgG
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR