Anti-HSP75/TRAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-1446R-FITC
BOSSBS-1446R-FITCEA 844 GBP
BOSSBS-1446R-FITC
Anti-HSP75/TRAP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antibodies
Chaperone that expresses an ATPase activity. Involved in maintaining mitochondrial function and polarization, most likely through stabilisation of mitochondrial complex I. Is a negative regulator of mitochondrial respiration able to modulate the balance between oxidative phosphorylation and aerobic glycolysis. The impact of TRAP1 on mitochondrial respiration is probably mediated by modulation of mitochondrial SRC and inhibition of SDHA.

Type: Primary
Antigen: HSP75/TRAP1
Clonality: Polyclonal
Clone:
Conjugation: FITC
Public Immunogen Range: 501-650/704
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR