Anti-CEP70 Rabbit Polyclonal Antibody (Alexa Fluor® 750)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13860R-A750
BOSSBS-13860R-A750EA 835 GBP
BOSSBS-13860R-A750
Anti-CEP70 Rabbit Polyclonal Antibody (Alexa Fluor® 750)
Antibodies
Plays a role in the organization of both preexisting and nascent microtubules in interphase cells. During mitosis, required for the organization and orientation of the mitotic spindle.

Type: Primary
Antigen: CEP70
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 750
Public Immunogen Range: 351-450/597
Host: Rabbit
Isotype: IgG
Reactivity: Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR