Anti-EPRS Rabbit Polyclonal Antibody
Supplier: Bioss
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
BS-13393R
BOSSBS-13393REA 680 GBP
BOSSBS-13393R
Anti-EPRS Rabbit Polyclonal Antibody
Antibodies
The fidelity of protein synthesis requires efficient discrimination of amino acid substrates by aminoacyl-tRNA synthetases. Aminoacyl-tRNA synthetases function to catalyze the aminoacylation of tRNAs by their corresponding amino acids, thus linking amino acids with tRNA-contained nucleotide triplets. ProRS (Prolyl-tRNA synthetase), also known as EPRS, EARS, PARS, QARS, QPRS, PIG32 or GLUPRORS, is a 1,512 amino acid protein that contains three WHEP-TRS domains and belongs to both the class-I and class-II aminoacyl-tRNA synthetase family. Functioning as a component of the multisynthase complex,More Product Information