Anti-Giantin/GOLGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)

Supplier: Bioss
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

BS-13356R-A680
BOSSBS-13356R-A680EA 835 GBP
BOSSBS-13356R-A680
Anti-Giantin/GOLGB1 Rabbit Polyclonal Antibody (Alexa Fluor® 680)
Antibodies
May participate in forming intercisternal cross-bridges of the Golgi complex.

Type: Primary
Antigen: Giantin/GOLGB1
Clonality: Polyclonal
Clone:
Conjugation: ALEXA FLUOR® 680
Public Immunogen Range: 751-850/3259
Host: Rabbit
Isotype: IgG
Reactivity: Human; Mouse; Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR